Waterproof Cryo Gloves - Shoulder Length
Waterproof Cryo Gloves - Shoulder Length
Actual product may vary
Thermo Fisher Scientific
Waterproof Cryo Gloves - Shoulder Length
Safely handle samples from ultra low temperature freezers and cryogenic storage with Thermo Scientific™ Waterproof Cryo Gloves. Available in elbow, mid-arm, and shoulder-length options.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
189448Medium
189449Large
Catalog number 189448
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
Medium
Safely handle samples from ultra low temperature freezers and cryogenic storage with Thermo Scientific™ Waterproof Cryo Gloves. Available in elbow, mid-arm, and shoulder-length options.
  • Provides splash protection
  • Comfortable, lightweight, breathable
  • Side release buckles for quick on/off
Specifications
DescriptionShoulder Length
Product TypeWaterproof Cryogenic Gloves
SizeMedium
For Use With (Application)Cryogenic Preservation
Length (English)28 in.
Length (Metric)71 cm
Unit Size