Waterproof Cryo Gloves - Elbow Length
Waterproof Cryo Gloves - Elbow Length
Actual product may vary
Thermo Fisher Scientific
Waterproof Cryo Gloves - Elbow Length
Safely handle samples from ultra low temperature freezers and cryogenic storage with Thermo Scientific™ Waterproof Cryo Gloves. Available in elbow, mid-arm, and shoulder-length options.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
189446
also known as PPXT-49
Large
189445Medium
189447X-Large
Catalog number 189446
also known as PPXT-49
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
Large
Safely handle samples from ultra low temperature freezers and cryogenic storage with Thermo Scientific™ Waterproof Cryo Gloves. Available in elbow, mid-arm, and shoulder-length options.
  • Provides splash protection
  • Comfortable, lightweight, breathable
  • Side release buckles for quick on/off
Specifications
DescriptionElbow Length
Product TypeWaterproof Cryogenic Gloves
SizeLarge
For Use With (Application)Cryogenic Preservation
Length (English)18 in.
Length (Metric)45.7 cm
Unit Size384 TESTS