Disposable Protective Overshoe Covers 442
Disposable Protective Overshoe Covers 442
Actual product may vary
Thermo Fisher Scientific
Disposable Protective Overshoe Covers 442
The 3M™ Disposable Overshoe Covers 442 are disposable shoe covers designed to provide basic barrier protection to help protect footwear against light liquid splashes and hazardous dusts.
Have Questions?
Catalog number 189442
View Price:Sign in to see your account pricing. Need an account? Register with us today.
A reliable, disposable boot cover designed to help protect footwear against light liquid splashes and hazardous dusts. Made from a microporous laminate material with an anti-static treatment.
  • Covers are elasticated to help ensure a good fit
  • Antistatic treatment
  • Universal size
Specifications
SizeUniversal
MaterialMicroporous Fabric
CoatingMicroporous Film Laminate
ColorWhite
DisposableSingle-use
Closure TypeElastic Ankle
Model442
ResistanceDry Particulates, Fluid
Product TypeBoot Covers
Unit Size