Chest Freezer Racks
Chest Freezer Racks
Actual product may vary
Chest Freezer Racks
Chest Freezer Racks
Chest Freezer Racks
Chest Freezer Racks
Chest Freezer Racks
Chest Freezer Racks
Chest Freezer Racks
Use Thermo Scientific™ Chest Freezer Racks with Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)Holds
189001Wire Basket Rail Set12.7cu. ft. freezersGeneral Freezer Items
3981842 in. Box Rack - Side Access Rack (Tower)3 cu. ft. chest freezersBoxes (2 in.)
3981862 in. Box Rack - Side Access Rack (Tower)13, 17, and 20 cu. ft. freezersBoxes (2 in.)
Catalog number 189001
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Wire Basket Rail Set
For Use With (Equipment):
12.7cu. ft. freezers
Holds:
General Freezer Items
Use Thermo Scientific™ Chest Freezer Racks with TDE Series Ultra-Low Temperature Chest Freezers as well as legacy Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
  • Warranty varies by country, please contact us for details
Specifications
Product TypeWire Basket (Rail Set)
DescriptionWire Basket Rail Set
ColorSilver
HoldsGeneral Freezer Items
ShapeRectangular
For Use With (Equipment)12.7cu. ft. freezers
Unit Size