384 Well Black Plate, TC Surface, Pack of 10
384 Well Black Plate, TC Surface, Pack of 10
Actual product may vary
384 Well Black Plate, TC Surface, Pack of 10
Color:Black; Sterility:Sterile; Lid:With Lid
Have Questions?
Catalog NumberdimFormatLengthMetric
164564
Catalog number 164564
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Improve background, autofluorescence and crosstalk with Thermo Scientific™ Nunc™ 384-Well Polystyrene Black and White Microplates.
  • Black microplates are recommended for fluorescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
LidWith Lid
Culture Area0.084 cm2/well
Array16 x 24
Packaging10 Pk., 30 Cases
Product LineNunc
ColorBlack
Product TypeMicroplate
MaterialPolystyrene
Volume (Metric) Well120 μL
No. of Wells384
Volume (Metric) Working10 to 100 μL
SterilitySterile
Surface TreatmentCell Culture
Well ShapeFlat Bottom (F-well)
No. per Case30
No. per Pack10
Unit Size