Work Area Covers for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Work Area Covers for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Actual product may vary
Thermo Fisher Scientific
Work Area Covers for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Work Area Covers for Thermo Scientific™ Refrigerated and Heated Bath Circulators include stainless-steel covers, and covers with leveling devices.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
160-0040Water Bath Model S49
160-0038Water Bath Models S21, S30, S45
160-0042Water Bath Model A10B
Catalog number 160-0040
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
Water Bath Model S49
Work Area Covers for Thermo Scientific™ Refrigerated and Heated Bath Circulators include stainless-steel covers, and covers with leveling devices.
  • Warranty varies by country, please contact us for details
Specifications
DescriptionStainless-steel work area cover
For Use With (Equipment)Water Bath Model S49
TypeCover
Unit Size