Nalgene™ Analytical Test Filter Funnels, 250mL, white with black grid
Actual product may vary
Nalgene™ Analytical Test Filter Funnels, 250mL, white with black grid
Perform microbiological QC testing and analysis of water, food/beverage, raw material and finished product with these economical sterile disposable funnels.
Have Questions?
Catalog Number
Pore Size
Capacity (Metric)
Color
145-2045
0.45 μm
250 mL
White with Black Grid
Catalog number 145-2045
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Pore Size:
0.45 μm
Capacity (Metric):
250 mL
Color:
White with Black Grid
Use these economical sterile disposable filter funnels to perform microbiological QC testing and analysis of water, food/beverage, raw material and finished product. The squeeze and twist funnel is easy to remove for retrieval of the CN membrane.
Features:
High impact polystyrene collars are rugged for reliable performance
Preassembled, sterile, individually are bagged to ensure sample integrity
Adapters accommodate No. 8 stoppers for use with 0.5 in. hole in vacuum manifolds or filter flasks
47mm diameter cellulose nitrate membrane is certified for water quality work
250mL sizes are useful when testing beverages that may foam
Cat. No. 147-0045 features gray membrane that turns black when wet, making it easier to detect and count growing colonies
Note: Designed for single use only
Includes: Six funnel adapters with each case
Specifications
Capacity (Metric)250 mL
Certifications/ComplianceMeet requirements of APHA Standard Methods and EPA requirements for water quality work.
ColorWhite with Black Grid
Diameter (Metric) Filter47 mm
For Use With (Equipment)Filter Manifold, Filtering Flask