Perform microbiological QC testing and analysis of water, food/beverage, raw material and finished product with these economical sterile disposable funnels.
Have Questions?
Change view
Catalog Number
Pore Size
Capacity (Metric)
Color
140-4045
0.45 μm
150 mL
White with Gray Grid
147-2045
0.45 μm
250 mL
Gray with Black Grid
2 Options
Catalog number 140-4045
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Pore Size:
0.45 μm
Capacity (Metric):
150 mL
Color:
White with Gray Grid
Use these economical sterile disposable filter funnels to perform microbiological QC testing and analysis of water, food/beverage, raw material and finished product. The squeeze and twist funnel is easy to remove for retrieval of the CN membrane.
Features:
High impact polystyrene collars are rugged for reliable performance
Preassembled, sterile, individually bagged to ensure sample integrity
Adapters accommodate No. 8 stoppers for use with 0.5 in. hole in vacuum manifolds or filter flasks
47mm diameter cellulose nitrate membrane is certified for water quality work
250mL sizes are useful when testing beverages that may foam
Cat. No. 147-0045 features gray membrane that turns black when wet, making it easier to detect and count growing colonies
Note: Designed for single use only
Includes: Six funnel adapters with each case
Specifications
Capacity (Metric)150 mL
Certifications/ComplianceMembrane conforms to APHA Standard Methods, 16th ed., Section 909A, B, C. Meets EPA requirements for water quality work.
ColorWhite with Gray Grid
DescriptionDia.: 47 mm; Pore: 0.45 μm; White with gray grid
Diameter (Metric) Filter47 mm
For Use With (Equipment)Filter Manifold, Filtering Flask