Fixed Volume Labofix Micropipettor
Fixed Volume Labofix Micropipettor
Actual product may vary
Thermo Fisher Scientific
Fixed Volume Labofix Micropipettor
Designed to meet the increased demand for fixed volume micropipettes that combine a good quality level with a certainly interesting price. Kartell™ Fixed Volume Labofix Micropipettor is ideal for applications in which a fixed dosage carried out with good precision and accuracy is necessary.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Compatible Tips
137350 μLKartell™ 958/960
13781000 μLKartell™ 961
1376250 μLKartell™ 961
Catalog number 1373
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
50 μL
Compatible Tips:
Kartell™ 958/960
  • Made of high quality plastic
  • With tip ejector and stainless steel piston for prolonged use over time
  • Handle connection ensures a more comfortable and non-fatiguing grip
  • Tip holder cone can house a wide variety of tips
  • It has a fine shape to make getting inside test tubes or containers having particularly narrow openings or lengthened shapes easier
  • Robust structure of the body ensures high mechanical and chemical resistances
Specifications
Accuracy±0.3%
Certifications/ComplianceEC/IVD, ISO8655
Compatible TipsKartell™ 958/960
AutoclavableAutoclavable
MaterialPlastic
ModelLabofix 50
TypeFixed Volume Micropipettor
Volume (Metric)50 μL
Precision±0.3%
Unit Size10 X 100 W