Nalgene™ Polypropylene Griffin Low-Form Beakers, 100mL, 100mL
Nalgene™ Polypropylene Griffin Low-Form Beakers, 100mL, 100mL
Actual product may vary
Nalgene™ Polypropylene Griffin Low-Form Beakers, 100mL, 100mL
Nalgene™ Polypropylene Griffin Low-Form Beakers, 100mL, 100mL
Nalgene™ Polypropylene Griffin Low-Form Beakers, 100mL, 100mL
Nalgene™ Polypropylene Griffin Low-Form Beakers, 100mL, 100mL
Nalgene™ Polypropylene Griffin Low-Form Beakers, 100mL, 100mL
Carry out precise and efficient pouring with these low-form beakers, featuring a single-spout design and easy-to-read silk-screened graduations.
Have Questions?
Catalog number 1201-0100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Carry out precise and efficient pouring with Thermo Scientific™ Nalgene™ Polypropylene Griffin Low-Form Beakers, featuring a single-spout design and easy-to-read silk-screened graduations.
Features:
  • Ribs for easy stacking
  • Silk-screened with graduations, size code, resin code, catalog number, max. temperature and “No Flame” symbol for easy identification and safe use
  • Autoclavable/Graduated
  • Meets ISO/DIN 7056 International Standards for Laboratory Plasticware Accuracy
Note:
Not for use on hot plates
Specifications
ColorNatural translucent
DescriptionBeaker Griffin Low-Form PP 100 mL
Capacity (Metric)100 mL
MaterialPPCO
Graduation Range10 to 100 mL
Increments10 mL
Quantity12/Pk.; 48/Cs.
TypeBeaker
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Certifications/ComplianceISO/DIN 7056
Scale10mL to 100mL in 10mL increments
StyleGriffin, Low Form
ACT LabelYes
Unit Size48