gamma-Butyrolactone, 99+%
gamma-Butyrolactone, 99+%
gamma-Butyrolactone, 99+%
gamma-Butyrolactone, 99+%
Acros Organics
gamma-Butyrolactone, 99+%
CAS: 96-48-0 | C4H6O2 | 86.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
10813010010 LPlastic drum
Catalog number 108130100
Price (MYR)
203.25
10LT
Add to cart
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
203.25
10LT
Add to cart
Chemical Identifiers
CAS96-48-0
IUPAC Nameoxolan-2-one
Molecular FormulaC4H6O2
InChI KeyYEJRWHAVMIAJKC-UHFFFAOYSA-N
SMILESO=C1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<40 APHA
Refractive index1.4350 to 1.4375 (20°C, 589 nm)
Infrared spectrumConforms
Water=<0.1 % (K.F.)
Appearance (Form)Clear liquid