Search Thermo Fisher Scientific
| Catalog Number | dimType | dimChannels | Sterility |
|---|---|---|---|
| 1064-15-6 | Sterile |
Handle reagents on automated systems simply and economically with Thermo Scientific™ Automation Reservoirs, which provide standard SBS dimensions ensuring compatibility with plate stackers and handlers. Reservoirs are disposable with no need for washing, allowing more reservoirs to be used per run. Built-in grooves in reservoir bottom reduce reagent waste and allow consistent aspiration.
Removable Splash Baffles: