TH-660 Swinging Bucket Rotor Package
TH-660 Swinging Bucket Rotor Package
Actual product may vary
Thermo Fisher Scientific
TH-660 Swinging Bucket Rotor Package
Process samples effectively with the TH-660 Swinging Bucket Rotor.
Have Questions?
Catalog NumberCapacity
105026 x 4.4 mL
Catalog number 10502
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
6 x 4.4 mL
Process samples effectively with the Thermo Scientific™ TH-660 Swinging Bucket Rotor Package.
Ordering Information:
Order Re-Seal 11 × 32mm tube vise (45537) separately. Contact your Sales Representative for information.
Compatible with:
Thermo Scientific Sorvall WX80/90/100 Ultra Centrifuge Series
Recommended for:
Rate zonal isolation of proteins and RNA.
Note:
Conical Adapter Removal Tool (Parts only available directly from Seton Scientific) can be ordered as 4209
Specifications
DescriptionTH-660 Swinging Bucket Rotor Package
MaterialTitanium
TypeSwinging Bucket Rotor
Capacity6 x 4.4 mL
Max. RCF488,576 x g
Max. Speed60,000 rpm
Radius (Metric)121.5 mm
Radius (Metric) Min.64.6 mm
K Factor44.4
Max. Tube Length60 mm
Max. Tube Diameter11 mm
Net Weight12.8 lb. (5.8 kg)
For Use With (Equipment)Thermo Scientific™ Sorvall™ WX ultraCentrifuges
Angle90°
Avg. RCF374,173 x g
Min. RCF259,770 x g
Unit Size