Fiberlite F50L-24 x 1.5 for WX+ Ultraspeed Centrifuges
Fiberlite F50L-24 x 1.5 for WX+ Ultraspeed Centrifuges
Actual product may vary
Thermo Fisher Scientific
Fiberlite F50L-24 x 1.5 for WX+ Ultraspeed Centrifuges
Achieve high maximum RCF and 14-place capacity with the Thermo Scientific™ Fiberlite™ F50L-24 x 1.5mL Fixed-Angle Rotor.
Have Questions?
Catalog NumberCapacity
096-24702824 x 1.5 mL
Catalog number 096-247028
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
24 x 1.5 mL
Process samples with high maximum RCF and 24-place capacity using the Thermo Scientific™ Fiberlite™ F50L-24 x 1.5mL Fixed-Angle Rotor.
Fiberlite rotors:
  • Reduce run-times with higher G-forces with our lightweight carbon fiber rotors
  • Ensure structural integrity with corrosion- and fatigue-resistant design
  • Backed by a 15-year warranty
Ordering information:
Tube adapters are sold separately, accommodating a variety of tubes varying in different capacities; available in packages of two. Contact your sales representative for more information.
Compatible with:
Thermo Scientific Sorvall™ WX, Sorvall Discovery™ SE Series and Beckman™ Optima L Series centrifuges, Nalgene™ bottles and tubes
Recommended for:
Small capacity density gradient centrifugation of sub cellular particles, nucleic acids and differential centrifugation of small particles.
Specifications
DescriptionFiberlite F50L-24 x 1.5 Fixed-Angle Rotor
MaterialCarbon Fiber Composite
TypeFixed Angle Rotor
Capacity24 x 1.5 mL
Max. RCF280,000 x g
Max. Speed50,000 rpm
Radius (Metric)100 mm
Radius (Metric) Min.72 mm
K Factor33
Max. Tube Length40 mm
Max. Tube Diameter11 mm
Net Weight11.5 lbs. (5.3 kg)
For Use With (Equipment)Thermo Scientific™ Sorvall™ WX ultraCentrifuges
Angle45°
Min. RCF201,600 x g
Unit Size