Barnstead™ Water System Cartridges
Barnstead™ Water System Cartridges
Actual product may vary
Barnstead™ Water System Cartridges
Barnstead™ Water System Cartridges
Thermo Fisher Scientific
Barnstead™ Water System Cartridges
Maintain your Thermo Scientific™ Barnstead™ point of use water purification system with these replacement cartridges.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
09.1006Ultrapure polisher cartridgeMicroPure
09.4011High purity water cartridge
09.1020Ultrapure polisher cartridgeSmart2Pure
09.2005Ultrapure polisher cartridgeGenPure GenPure Pro, GenPure xCAD, LabTower EDI
Catalog number 09.1006
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Ultrapure polisher cartridge
For Use With (Equipment):
MicroPure
Maintain your Thermo Scientific™ Barnstead™ point of use water purification systems with replacement Thermo Scientific™ Barnstead™ Cartridges.
Choose the appropriate part number for your system. These cartridges are for use with the following Barnstead point of use water purification systems:
  • MicroPure™
  • GenPure™
  • GenPure™ Pro
  • GenPure™ xCAD
  • Smart2Pure™
  • LabTower™ EDI
  • Pacific™ TII
Specifications
DescriptionUltrapure polisher cartridge
For Use With (Equipment)MicroPure
TypeCartridge, Ultrapure Polisher
Unit Size