Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps
Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps
Actual product may vary
Thermo Fisher Scientific
Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps
Maintain UV function with Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
09.1002MicroPure, Smart2Pure (UV and UV/UVF only)
09.4002Pacific TII; LabTower TII
50139226Pacific TII; LabTower TII
09.2002GenPure (UV, UV/UF, UV-TOC and UF/UV-TOC only), LabTower EDI
Catalog number 09.1002
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
MicroPure, Smart2Pure (UV and UV/UVF only)
Maintain UV function with Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Replacement UV Lamps, for use with Barnstead point of use water purification systems.
Ideal for use with the following Barnstead point of use water purification systems:
09.1002:
  • MicroPure™ (UV and UV/UVF systems only)
  • Smart2Pure™ (UV and UV/UVF systems only)
09.2002:
  • GenPure™ (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
  • GenPure™ Pro (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
  • GenPure™ xCAD (UV, UV/UF, UV-TOC and UF/UV-TOC systems only)
  • LabTower™ EDI
09.4002:
  • Pacific™ TII
  • LabTower™ TII
09.5002:
For storage reservoirs (230V only) used with the following Barnstead water purification systems:
  • LabTower RO
  • LabTower TII
  • Pacific TII (30 L, 60 L, 100 L only)
  • Smart2Pure (12 only)
Specifications
DescriptionReplacement UV lamp
For Use With (Equipment)MicroPure, Smart2Pure (UV and UV/UVF only)
TypeUV Lamp
Unit Size