Point of Use Filter Inserts
Point of Use Filter Inserts
Actual product may vary
Thermo Fisher Scientific
Point of Use Filter Inserts
Protect installations and systems from undissolved substances in water with these pre-filters, final filters and sterile filters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFlow RateFor Use With (Equipment)
06.5101Prefilter 1µm (10“)1000 L/hr. (at D p = 0.2 bar)Smart2Pure
06.5201Prefilter 5µm+activated carbon (10“)750 L/hr. (at D p = 0.2 bar)LabTower EDI, LabTower RO
Catalog number 06.5101
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Prefilter 1µm (10“)
Flow Rate:
1000 L/hr. (at D p = 0.2 bar)
For Use With (Equipment):
Smart2Pure
Protect installations and systems from undissolved substances in water with these pre-filters, final filters and sterile filters. Thermo Scientific™ Point of Use Filter Inserts include 10- and 20-inch pre-filters, final filters and sterile filters for Point of Use systems.
Pre-filters 1μm, 5μm
  • Protect installations and systems from undissolved substances in water
Final filters
  • Protect systems from undissolved substances in water
  • Usually used for final filtration
Sterile filters
  • For final filtration and delivery of organic-free water
Specifications
Cartridge Size10 in.
DescriptionPrefilter 1µm (10“)
For Use With (Equipment)Smart2Pure
TypePrefilter Insert
Flow Rate1000 L/hr. (at D p = 0.2 bar)
Pore Size (Metric)1 μm
Unit Size